Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-2 Hamster anti-Mouse, Human, Alexa Fluor™ 488, Clone: 3F11, BD Phrmingen™
Hamster Monoclonal Antibody
$88.00 - $192.00
Specifications
Antigen | Bcl-2 |
---|---|
Clone | 3F11 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry, Control |
Classification | Monoclonal |
Description
Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Mouse|Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
Bcl-2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
12043 | |
IgG1 | |
Affinity Chromatography |
3F11 | |
Flow Cytometry, Control | |
Alexa Fluor 488 | |
Hamster | |
Mouse, Human | |
B cell leukemia/lymphoma 2; Bcl-2; Bcl2; apoptosis regulator Bcl-2 | |
Recombinant Mouse Bcl-2 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title