Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Mouse anti-Human, AlexaFluor™488, Clone: AW604, MilliporeSigma™

Catalog No. 05826AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05826AF488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05826AF488 Supplier MilliporeSigma Supplier No. 05826AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Bcl-2 Antibody, clone AW604, Alexa Fluor™ 488 Conjugate is validated for use in Immunocytochemistry, Flow Cytometry for the detection of Bcl-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2
Applications Immunocytochemistry, Flow Cytometry
Classification Monoclonal
Clone AW604
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 488
Gene Symbols BCL2
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 41-54 (C-GAAPAPGIFSSQPG) of human Bcl-2.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_000624
Test Specificity Expected to react with both Alpha and Beta spliced isoforms reported by UniProt (P10415-1 and P10415-2, respectively).
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.