Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-2 related protein A1 Antibody (OTI3D10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246568
Description
Ensure accurate, reproducible results in Western Blot
Bcl-2 related protein A1 Monoclonal specifically detects Bcl-2 related protein A1 in Human samples. It is validated for Western Blot.Specifications
Bcl-2 related protein A1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 | |
ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS | |
Mouse | |
20 kDa | |
0.1 mL | |
Apoptosis, Cancer | |
597 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI3D10 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
BCL2A1 | |
Full length human recombinant protein of human BCL2A1 (NP_004040) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction