Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-3 Monoclonal Antibody (150-3.5), QED Bioscience™
SDP

Catalog No. 502277379
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7379 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7379 Supplier Leinco Technologies Inc Supplier No. 11067100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hamster Monoclonal Antibody

Bcl-3 Monoclonal antibody specifically detects Bcl-3 in Human, Mouse samples. It is validated for ELISA, Western Blot

Specifications

Antigen Bcl-3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 150-3.5
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Bcl3
Gene Accession No. P20749, Q9Z2F6
Gene Alias AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3
Host Species Hamster
Immunogen Recombinant mouse Bcl-3-6His.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12051, 602
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.