Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bcl-3 Monoclonal Antibody (Ham 150-3.5), FITC
GREENER_CHOICE

Catalog No. PIMA517667
Change view
Click to view available options
Quantity:
100 μg
300 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517667 100 μg
PIMA517668 300 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA517667 Supplier Invitrogen™ Supplier No. MA517667
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Bcl-3 Monoclonal Antibody for Flow

B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-3
Applications Flow Cytometry
Classification Monoclonal
Clone Ham 150-3.5
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Bcl3
Gene Accession No. Q9Z2F6
Gene Alias AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3
Gene Symbols Bcl3
Host Species Armenian Hamster
Immunogen recombinant Bcl-3-6-histidine tag.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12051
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.