Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bcl-3 Monoclonal Antibody (Ham150-3.5)
GREENER_CHOICE

Catalog No. MA1950
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1-950 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1-950 Supplier Invitrogen™ Supplier No. MA1950
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

MA1-950 detects human and mouse Bcl-3 from transfected cells and shows no cross-reactivity to other Bcl isoforms. MA1-950 has been successfully used in Western blot and FACS procedures. By Western blot, this antibody detects an ~47 kDa protein representing Bcl-3 from C57BI/6 human T cells and Bcl-3 transgenic ms T cells. MA1-950 immunogen is full length Bcl-3 protein from mouse.

B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-3
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone Ham150-3.5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Bcl3
Gene Accession No. P20749, Q9Z2F6
Gene Alias AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3
Gene Symbols Bcl3
Host Species Armenian Hamster
Immunogen Full length mouse Bcl-3 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12051, 602
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.