Learn More
Description
Host Species: Mouse
Clone: BL6.02, same as PG-B6p
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Rabbit, Rat, Sheep and Pig. Others not tested.
Epitope: aa 3-484
Immunogen: Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein
Positive Control: Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Bcl-6 Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | BL6.02; same as PG-B6p |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein |
| Quantity | 12 mL |
| Regulatory Status | RUO |
| Research Discipline | Apoptosis |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.