Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Bcl-6 Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1114S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1114S0 100 μL Concentrated
MS1114S1 500 μL Concentrated
MS1114S 1 mL Concentrated
MS1114RQ 12 mL Ready-to-Use
MS1114R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1114S0 Supplier Epredia™ Supplier No. MS1114S0

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Bcl-6 Ab-2, Mouse Monoclonal Antibody.

Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin's lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt's lymphomas and in nodular, lymphocyte predominant Hodgkin's disease.

Host Species: Mouse

Clone: BL6.02, same as PG-B6p

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Rabbit, Rat, Sheep and Pig. Others not tested.

Epitope: aa 3-484

Immunogen: Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Bcl-6 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BL6.02; same as PG-B6p
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 3-484 of the human Bcl-6 protein
Purification Method Tonsil
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Bovine, Human, Pig, Rabbit, Rat, Sheep
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.