Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-6 Antibody (rBCL6/1475), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308835AF405
Description
Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human samples. It is validated for Western BlotSpecifications
Bcl-6 | |
Monoclonal | |
Alexa Fluor 405 | |
B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S | |
Recombinant human bcl-6 protein fragment (around aa256-389) (exact sequence is proprietary) (Uniprot: P41182) | |
100 μg | |
Cancer, Cell Cycle and Replication, Tumor Suppressors | |
604 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot | |
rBCL6/1475 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction