Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-6 Mouse anti-Human, Mouse, BUV 496, Clone: K112-91, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567753
This item is not returnable.
View return policy
Description
Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human, Mouse samples. It is validated for Intracellular staining (Flow Cytometry)
Specifications
Bcl-6 | |
Monoclonal | |
Brilliant Ultraviolet 496 | |
BCL6; B-cell lymphoma 6 protein; LAZ3; Laz-3, ZBTB27, ZNF51 | |
Human Bcl-6 Recombinant Protein | |
RUO | |
Human, Mouse | |
Liquid |
Flow Cytometry | |
K112-91 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction