Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-6 Mouse anti-Human, Mouse, BV480, Clone: K112-91, BD Horizon™
Mouse Monoclonal Antibody
$261.50 - $446.00
Specifications
Antigen | Bcl-6 |
---|---|
Clone | K112-91 |
Applications | Flow Cytometry, Control, Immunofluorescence, Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Conjugate | Brilliant Violet 480 |
Description
Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)|Immunofluorescence|Immunohistochemistry-frozenSpecifications
Bcl-6 | |
Flow Cytometry, Control, Immunofluorescence, Immunohistochemistry (Frozen) | |
Brilliant Violet 480 | |
Mouse | |
Human, Mouse | |
BCL6; B-cell lymphoma 6 protein; LAZ3; Laz-3, ZBTB27, ZNF51 | |
IgG1 κ | |
Affinity Chromatography |
K112-91 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Bcl-6 Recombinant Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title