Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL-6 Polyclonal Antibody, Abbomax

Catalog No. 502204015
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4015 Supplier Abbomax Inc Supplier No. 601840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Lymphoma

BCL-6 Polyclonal antibody specifically detects BCL-6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen BCL-6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene BCL6
Gene Accession No. P41182
Gene Alias B cell CLL/lymphoma 6; B cell leukemia/lymphoma 6; B-cell CLL/lymphoma 6; B-cell CLL/lymphoma 6 (zinc finger protein 51); B-cell leukemia/lymphoma 5; B-cell leukemia/lymphoma 6; B-cell lymphoma 5 protein; B-cell lymphoma 6 protein; B-cell lymphoma 6 protein homolog; B-cell lymphoma 6 protein transcript; BCL5; BCL-5; Bcl6; Bcl-6; BCL6 transcription repressor; BCL6A; cys-his2 zinc finger transcription factor; LAZ3; LAZ-3 protein; lymphoma-associated zinc finger gene on chromosome 3; protein LAZ-3; ZBTB27; zinc finger and BTB domain-containing protein 27; Zinc finger protein 51; zinc finger transcription factor BCL6S; ZNF51
Host Species Rabbit
Immunogen A synthetic peptide corresponding to inter-segment of human BCL-6 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 604
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.