Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bcl-B Monoclonal Antibody (8A11G12)
GREENER_CHOICE

Catalog No. PIMA531648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531648 Supplier Invitrogen™ Supplier No. MA531648
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31648 has been tested in indirect ELISA.

The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains conserved BH4, BH1 and BH2 domains. This protein can interact with other members of BCL-2 protein family including BCL2, BCL2L1/BCL-X(L), and BAX. Overexpression of this gene has been shown to suppress cell apoptosis possibly through the prevention of cytochrome C release from the mitochondria, and thus activating caspase-3 activation. The mouse counterpart of this protein is found to interact with Apaf1 and forms a protein complex with Caspase 9, which suggests the involvement of this protein in APAF1 and CASPASE 9 related apoptotic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-B
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 8A11G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene BCL2L10
Gene Accession No. Q9HD36
Gene Alias AA420380; anti-apoptotic protein Boo; Anti-apoptotic protein NrH; apoptosis regulator Bcl-B; AU023065; Bcl-2 homolog Diva; BCL2 like 10; BCL2L10; bcl2-L-10; Bcl2-like 10; BCL2-like 10 (apoptosis facilitator); bcl-2-like protein 10; BCLB; BCL-B; Boo; C85687; death inducer binding to vBcl-2 and Apaf-1; Diva
Gene Symbols BCL2L10
Host Species Mouse
Immunogen Purified recombinant fragment of human BCL2L10 (AA: 31-186) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10017
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.