Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl G Antibody (OTI6D1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272204B
Description
Bcl G Monoclonal specifically detects Bcl G in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Bcl G | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
BCL2L14 | |
Recombinant protein fragment corresponding to amino acids 116-216 of human Bcl G (NP_620048) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI6D1 | |
Western Blot, Immunohistochemistry | |
apoptosis facilitator Bcl-2-like protein 14, Apoptosis regulator Bcl-G, Bcl2-L-14, BCL2-like 14 (apoptosis facilitator), BCL-G, BCLGbcl2-L-14 | |
Mouse | |
Affinity Purified | |
Apoptosis | |
79370.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction