Learn More
Invitrogen™ Bcl-G Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA14209
Description
PA1-4209 detects the long form of BCL-G in in human samples. PA1-4209 has been successfully used in Western blot procedures. The PA1-4209 immunogen is a synthetic peptide corresponding to residues T(298) K Y L K E N F S P W I Q Q H(312) of the long form of human Bcl-G.
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
Specifications
Bcl-G | |
Polyclonal | |
Unconjugated | |
BCL2L14 | |
4930452K23Rik; 4933405K19Rik; 9030625M01Rik; AI429211; Apoptosis facilitator Bcl-2-like protein 14; apoptosis regulator BCL-G; BCL2 like 14; Bcl2l14; bcl2-L-14; BCL2-like 14 (apoptosis facilitator); BCLG; testicular tissue protein Li 26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79370 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9BZR8 | |
BCL2L14 | |
Synthetic peptide corresponding to residues T(298) K Y L K E N F S P W I Q Q H(312) of the long form of human Bcl-G. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.