Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl rambo Antibody (Rocky-2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197578
Description
Bcl rambo Monoclonal specifically detects Bcl rambo in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
Bcl rambo | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:300, Immunoprecipitation 1:100 | |
BCL2L13 | |
Recombinant human Bcl-rambo. | |
0.05 mg | |
Apoptosis | |
23786 | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Rocky-2 | |
Unconjugated | |
Bcl2-L-13, BCL2-like 13 (apoptosis facilitator), bcl-2-like protein 13, Bcl-rambo, MIL1BCL-RAMBO, Protein Mil1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes the C-terminus of human Bcl-rambo (aa 224-459). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction