Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bcl-rambo Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111313 Supplier Invitrogen™ Supplier No. PA5111313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASSSTVPLG FHYETKYVVL SYLGLLSQEK LQEQHLSSPQ GVQLDIASQS LDQEILLKVK TEIEEELKSL DKEISEAFTS TGFDRHTS Highest antigen sequence indentity to the following orthologs: Rat - 78%, Mouse - 77%.

Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Members of the Bcl-2 family are known to be critical regulators of this process. These proteins are characterized by the presence of several conserved motifs termed Bcl-2 homology (BH) domains. A novel, widely expressed member termed Bcl-rambo was recently identified. This protein is localized to mitochondria in mammalian cells and its overexpression induces apoptosis which could be blocked by co-expression of inhibitor of apoptosis proteins (IAPs) such as XIAP, cIAP1, and cIAP2. Bcl-rambo shows overall homology to the anti-apoptotic members containing BH motifs, but unlike Bcl-2, the C-terminal membrane anchor of Bcl-rambo is preceded by a unique 250 amino acid insertion. This region by itself can induce apoptosis more efficiently than the Bcl-2 homology regions, suggesting that Bcl-rambo may be important other pro-apoptotic pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-rambo
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BCL2L13
Gene Accession No. Q9BXK5
Gene Alias BCL2 like 13; BCL2L13; Bcl2-L-13; BCL2-like 13 (apoptosis facilitator); bcl-2-like protein 13; Bcl-rambo; CD003; E430016C20Rik; MIL1; Mil-1; mitochondria located 1 homolog; protein Mil1
Gene Symbols BCL2L13
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Bcl-rambo. Recombinant protein control fragment (Product #RP-104897).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23786
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.