Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL-W/BCL2L2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298720JF549
Description
BCL-W/BCL2L2 Polyclonal antibody specifically detects BCL-W/BCL2L2 in Human samples. It is validated for ELISASpecifications
BCL-W/BCL2L2 | |
Polyclonal | |
ELISA | |
Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2-like 2, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2 | |
Produced in rabbits immunized with purified, recombinant Human BCL-W/BCL2L2 (Uniprot#: Q92843-1; Met 1-Thr 172). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer | |
599 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction