Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL-W/BCL2L2 Mouse anti-Human, Clone: CPTC-BCL2L2-2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30745620UG
Description
BCL-W/BCL2L2 Monoclonal specifically detects BCL-W/BCL2L2 in Human samples. It is validated for Western Blot, Protein Array.Specifications
BCL-W/BCL2L2 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Protein Array | |
Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2-like 2, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2 | |
Recombinant human full-length BCL-W/BCL2L2 protein | |
20 μg | |
Apoptosis, Biologically Active Proteins, Cancer | |
599 | |
Store at 4°C. | |
IgG1 κ |
Western Blot, Peptide Array | |
CPTC-BCL2L2-2 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction