Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bcl-W Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588153
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588153 Supplier Invitrogen™ Supplier No. PA588153
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549; Cellular Location: Mitochondrion membrane, Peripheral membrane protein Immunogen sequence: MATPASAPDT RALVADFVGY KLRQKGYVCG AGPGEGPAAD PLHQAMRAAG DEFETRFRRT FSDLAAQLHV TPGSAQQRFT QVSDELFQGG PNWGRLVAFF VFGAALCAES VNKEMEPLVG QVQEWMVAYL ETQLADWIHS SGGWAEFTAL YGDGALEEAR RLREGNWASV RTVLTGAVAL GALVTVGAFF ASK.

Using a PCR-based strategy, an additional protein with life-promoting activity, designated Bcl-w, has been identified. The protein is highly conserved between mouse and human and is encoded by a gene located near the TCRalpha gene on chromosome 14. Bcl-w is expressed in myeloid cell lines but not in T and B lymphocytes, and can be found in a wide range of tissues. An alternative splicing event in exon 4 results in two transcripts. The first, Bcl-w, encodes a protein of 193 amino acids, and the second, Bcl-w/rox, encodes a protein 333 amino acids in length. The 'rox' portion of Bcl-w/rox shows a striking 66% amino acid sequence identity with the Drosophila rox2 protein; however, the Bcl-w/rox transcript may be expressed at very low levels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-W
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene BCL2L2
Gene Accession No. P70345, Q92843
Gene Alias Apoptosis regulator Bcl-W; AW048834; BCL2 like 2; BCL2L2; BCL2-L-2; BCL2-like 2; BCL2-like 2 protein; Bcl-2-like protein 2; Bcl2-like protein 2; BCLW; BCL-W; BCL-WEL; BCL-WS; c98; Gtrgal2; Gtrosa41; Kiaa0271; PPP1R51; protein phosphatase 1, regulatory subunit 51
Gene Symbols BCL2L2
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12050, 599, 60434
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.