Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Bcl-w Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF824SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF824SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF824SP Supplier R&D Systems Supplier No. AF824SP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 2 publications

BCL-W/BCL2L2 Polyclonal specifically detects BCL-W/BCL2L2 in Human∣Mouse samples. It is validated for Western Blot.

Specifications

Antigen BCL-W/BCL2L2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1.2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q92843
Gene Alias Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2L2, BCL2-like 2, Bclw, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2
Gene Symbols BCL2L2
Host Species Rabbit
Immunogen KLH-coupled human/mouse Bcl-w synthetic peptide Gly32-Asp51 Accession # Q92843
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 599
Test Specificity Detects human and mouse Bcl-w in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.