Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
bcl-x Antibody (2H12), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234531AF350
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Bcl-xL | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
BCL2L1 | |
A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-xL protein (Uniprot: Q07817) | |
0.1 mL | |
Primary | |
Recognizes a protein of 27kDa, identified as the Bcl-X protein. This monoclonal antibody shows no cross-reaction with Bcl-2 or Bax protein. Bcl-X has two isoforms, Bcl-XL (long), a 241 amino acid protein which suppresses cell death. And Bcl-XS (short), a 178 amino acid protein lacking a 63 amino acid domain which functions as a dominant inhibitor of Bcl-2. This monoclonal antibody reacts with both Bcl-XS and Bcl-XL proteins. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2H12 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
Mouse | |
Protein A or G purified | |
Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
598.0 | |
Human, Mouse, Rat, Pig | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction