Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
bcl-x Antibody (BX006), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225240
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Bcl-xL | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 0.5-1 μg/mL, Flow Cytometry 0.5-1 μg/million cells, ELISA 1-5 μg/mL for coating, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunoprecipitation 1-2 μg/500 μg protein lysate, Immunohistochemistry-Paraffin 0.5-1 μg/mL, Immunohistochemistry-Frozen 0.5-1 μg/mL | |
Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
Mouse | |
27 kDa | |
0.1 mg | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
598 | |
Human, Mouse, Rat, Pig | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
BX006 | |
Unconjugated | |
NP_001182 | |
BCL2L1 | |
A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-xL protein. (Uniprot: Q07817) | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of 27kDa, identified as the Bcl-X protein. This monoclonal antibody shows no cross-reaction with Bcl-2 or Bax protein. Bcl-X has two isoforms, Bcl-XL (long), a 241 amino acid protein which suppresses cell death. And Bcl-XS (short), a 178 amino acid protein lacking a 63 amino acid domain which functions as a dominant inhibitor of Bcl-2. This monoclonal antibody reacts with both Bcl-XS and Bcl-XL proteins. | |
Store at 4C. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction