Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
bcl-x Antibody (SPM519), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308595V
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
bcl-x | |
Monoclonal | |
DyLight 405 | |
Apoptosis regulator Bcl-X;BCL2L;BCL2L1;Bcl-2-like protein 1;BCLX;Bcl-X;BCL-XL/S;PPP1R52 | |
Recombinant human bcl-x protein fragment (around aa 1-135) (exact sequence is proprietary) (Uniprot: Q07817) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
SPM519 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
598 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction