Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
bcl-x Mouse anti-Human, Clone: BCL2L1/2406, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30745520UG
Description
bcl-x Monoclonal specifically detects bcl-x in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
bcl-x | |
Monoclonal | |
0.2 mg/ml | |
ELISA 1-5 ug/ml, Immunohistochemistry-Paraffin | |
Mouse | |
Protein A or G purified | |
RUO | |
598 | |
Store at 4°C. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
BCL2L1/2406 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Recombinant human bcl-x protein fragment (around aa 1-135) (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction