Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
bcl-x Mouse anti-Mouse, Rat, Clone: 2H12, Novus Biologicals™

Mouse Monoclonal Antibody
$219.50 - $487.50
Specifications
Antigen | bcl-x |
---|---|
Clone | 2H12 |
Dilution | Western Blot 1 - 5 ug/ml |
Applications | Western Blot |
Classification | Monoclonal |
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Mouse, Rat samples. It is validated for Western BlotSpecifications
bcl-x | |
Western Blot 1 - 5 ug/ml | |
Monoclonal | |
Mouse | |
Mouse, Rat | |
598 | |
IgG2a | |
Store at -20C. Avoid freeze-thaw cycles. |
2H12 | |
Western Blot | |
Unconjugated | |
RUO | |
PBS | |
A synthetic peptide corresponding to amino acids 3 to 14 of the human bcl-xl sequence (Uniprot: Q07817) | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title