Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bcl-X Polyclonal Antibody

Catalog No. PIPA146479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA146479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA146479 Supplier Invitrogen™ Supplier No. PA146479

Rabbit Polyclonal Antibody

The target sequence has 93% sequence homology with porcine, 87% to mouse and 81% to rat. This antibody is not recommended for ICC/IF applications.

The protein encoded the BCL-X gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The proteins encoded by this gene are located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential, and thus controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Alternative splicing results in multiple transcript variants encoding two different isoforms. The longer isoform acts as an apoptotic inhibitor and the shorter isoform acts as an apoptotic activator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-X
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Phosphate, Tris citrate with 0.1% sodium azide; pH 7-8
Gene BCL2L1
Gene Accession No. Q07817
Gene Alias anti-apoptosis regulatory protein; anti-apoptotic Bcl-2 gene family member; anti-apoptotic Bcl-xL; anti-apoptotic regulator Bcl-xL; apoptosis regulator Bcl-X; B cell lymphoma 2 like; B cell lymphoma like X; B-cell leukemia/lymphoma x; Bcl 2 like 1; bcl x; Bcl xL; BCL XL/S; Bcl(X)L; bcl-2 L1; BCL2 like 1; BCL2L; Bcl2l1; bcl2-L-1; BCL2-like 1; Bcl-2-like protein 1; BCL2-like protein 1; BCLX; bcl-x; Bcl-x long protein; BclxL; Bcl-xL; BCL-XL/S; BclXS; bcl-xS; BLC2L; DKFZp781P2092; PPP1R52; protein phosphatase 1, regulatory subunit 52; RP5-857M17.3; unnamed protein product
Gene Symbols BCL2L1
Host Species Rabbit
Immunogen Synthetic peptide made to an N-terminal portion of human BCL2L1 (within residues 30-70). [Swiss-Prot# Q07817].
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403618, 443061, 493701, 598
Target Species Human, Canine, Sheep, Feline
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.