Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-X Polyclonal Antibody, Abbomax

Catalog No. 502203693
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3693 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3693 Supplier Abbomax Inc Supplier No. 5003374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Raji cells Cellular location: Cytoplasmic.

Bcl-X Polyclonal antibody specifically detects Bcl-X in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot

Specifications

Antigen Bcl-X
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene BCL2L1
Gene Accession No. Q07817, Q64373
Gene Alias anti-apoptosis regulatory protein; anti-apoptotic Bcl-2 gene family member; anti-apoptotic Bcl-xL; anti-apoptotic regulator Bcl-xL; apoptosis regulator Bcl-X; B cell lymphoma 2 like; B cell lymphoma like X; B-cell leukemia/lymphoma x; Bcl 2 like 1; bcl x; Bcl xL; BCL XL/S; Bcl(X)L; bcl-2 L1; BCL2 like 1; BCL2L; Bcl2l1; bcl2-L-1; BCL2-like 1; Bcl-2-like protein 1; BCL2-like protein 1; BCLX; bcl-x; Bcl-x long protein; BclxL; Bcl-xL; BCL-XL/S; BclXS; bcl-xS; BLC2L; DKFZp781P2092; PPP1R52; protein phosphatase 1, regulatory subunit 52; RP5-857M17.3; unnamed protein product
Host Species Rabbit
Immunogen A synthetic peptide derived from N-terminus of human BCL-X protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12048, 598
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.