Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-xL Antibody (016), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289507UV
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human samples. It is validated for Western Blot, ELISA.Specifications
Bcl-xL | |
Monoclonal | |
DyLight 350 | |
Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Bcl-xL (NP_612815.1; Met1-Arg212). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
598 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
16 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction