Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-xL Antibody (042), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290348B
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Mouse samples. It is validated for ELISA.Specifications
Bcl-xL | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
42 | |
ELISA | |
Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Bcl-xL (Q64373-1; Met1-Arg212). | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
598 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction