Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-xL Antibody (OTI4A9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270247FR
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Bcl-xL | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
BCL2L1 | |
Recombinant fragment expressed in E.coli corresponding to amino acids 1-212 of human Bcl-xL. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI4A9 | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
Mouse | |
Affinity Purified | |
Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
598.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction