Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-XL Mouse anti-Human, Clone: OTI4A9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA500097

Description
BCL-XL is a part of the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. BCL-XL is located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential and controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported. The longer isoform acts as an apoptotic inhibitor and the shorter form acts as an apoptotic activator.Specifications
Bcl-XL | |
Monoclonal | |
0.25 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q07817 | |
BCL2L1 | |
Recombit fragment expressed in E.coli corresponding to amino acids 1-212 of human Bcl-XL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4A9 | |
Unconjugated | |
BCL2L1 | |
BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52 | |
Mouse | |
Protein A | |
RUO | |
598 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction