Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL10 Monoclonal Antibody (OTI6A4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502233S


Description
BCL10 Monoclonal antibody specifically detects BCL10 in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
BCL10 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O15355 | |
Mouse | |
Affinity chromatography | |
RUO | |
475703, 5496 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI6A4 | |
Unconjugated | |
Ppm1g | |
AI132454; B cell CLL/lymphoma 10; B cell leukemia/lymphoma 10; B-cell CLL/lymphoma 10; B-cell leukemia/lymphoma 10; B-cell lymphoma/leukemia 10; BCL10; Bcl-10; BCL10 immune signaling adaptor; C81403; CARD containing molecule enhancing NF-kB; CARD-containing apoptotic signaling protein; CARD-containing molecule enhancing NF-kappa-B; CARD-containing proapoptotic protein; CARD-like apoptotic protein; CARMEN; caspase-recruiting domain-containing protein; cCARMEN; cE10; c-E10; CED-3/ICH-1 prodomain homologous E10-like regulator; cellular homolog of vCARMEN; Cellular-E10; CIPER; Clap; hCLAP; IMD37; Mamm; mammalian CARD-containing adapter molecule E10; mCIPER; mCLAP; mE10; OTTHUMP00000011647; OTTHUMP00000036080; RCD; RP11-234D19.2; R-RCD1 | |
Full length human recombinant protein of human PPM1G produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction