Learn More
BCL10 Mouse anti-Human, Clone: OTI4H9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800445

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.Specifications
BCL10 | |
Monoclonal | |
Unconjugated | |
BCL10 | |
CARMEN, CIPER, CLAP, IMD37, c-E10, mE10 | |
Mouse | |
Affinity Chromatography | |
RUO | |
8915 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4H9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95999 | |
BCL10 | |
Full length human recombit protein of human BCL10 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.