Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCL11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526635
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526635 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526635 Supplier Invitrogen™ Supplier No. PA526635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCL11A Polyclonal Antibody for Western Blot

This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL11A
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene BCL11A
Gene Accession No. Q9H165
Gene Alias 2810047E18Rik; B cell CLL/lymphoma 11A; B cell CLL/lymphoma 11A (zinc finger protein); BAF chromatin remodeling complex subunit BCL11A; B-cell CLL/lymphoma 11A; B-cell CLL/lymphoma 11A (zinc finger protein); B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2; B-cell lymphoma/leukemia 11A; BCL11A; BCL-11A; BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1; BCL11A, BAF complex component; BCL11A-L; BCL11a-M; BCL11A-S; BCL11A-XL; C2H2-type zinc finger protein; COUP-TF interacting protein 1; COUP-TF-interacting protein 1; CTIP1; D930021L15Rik; ecotropic viral integration site 9 homolog; ecotropic viral integration site 9 protein; Ecotropic viral integration site 9 protein homolog; Evi9; EVI-9; Evi9a; Evi9b; Evi9c; FLJ10173; FLJ34997; HBFQTL5; Kiaa1809; mKIAA1809; myeloid leukemia; zinc finger protein 856; ZNF856
Gene Symbols BCL11A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 806-835 amino acids from the C-terminal region of human BCL11A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53335
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.