Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BCL11A Recombinant Rabbit Monoclonal Antibody (BLR151J)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | BCL11A |
---|---|
Clone | BLR151J |
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544487
|
Invitrogen™
MA544487 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544488
|
Invitrogen™
MA544488 |
20 μL |
Each for $173.10
|
|
|||||
Description
BCL11A Recombinant Monoclonal Antibody for Western Blot, IP
This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
BCL11A | |
1 mg/mL | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 0.09% sodium azide; pH 6.8-7.4 | |
Q9H165 | |
53335 | |
Peptide representing a region between residue 785 and the C-terminus (residue 835) of human B-cell lymphoma/leukemia 11A. | |
Antibody |
BLR151J | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
BCL11A | |
2810047E18Rik; B cell CLL/lymphoma 11A; B cell CLL/lymphoma 11A (zinc finger protein); BAF chromatin remodeling complex subunit BCL11A; B-cell CLL/lymphoma 11A; B-cell CLL/lymphoma 11A (zinc finger protein); B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2; B-cell lymphoma/leukemia 11A; BCL11A; BCL-11A; BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1; BCL11A, BAF complex component; BCL11A-L; BCL11a-M; BCL11A-S; BCL11A-XL; C2H2-type zinc finger protein; COUP-TF interacting protein 1; COUP-TF-interacting protein 1; CTIP1; D930021L15Rik; ecotropic viral integration site 9 homolog; ecotropic viral integration site 9 protein; Ecotropic viral integration site 9 protein homolog; Evi9; EVI-9; Evi9a; Evi9b; Evi9c; FLJ10173; FLJ34997; HBFQTL5; Kiaa1809; mKIAA1809; myeloid leukemia; zinc finger protein 856; ZNF856 | |
BCL11A | |
Primary | |
purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title