Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL11B Antibody (1F8H9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | BCL11B |
---|---|
Clone | 1F8H9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61700
![]() |
Novus Biologicals
NBP261700 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
BCL11B Monoclonal specifically detects BCL11B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
BCL11B | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
B-cell CLL/lymphoma 11B, B-cell CLL/lymphoma 11B (zinc finger protein), B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein, B-cell lymphoma/leukaemia 11B, B-cell lymphoma/leukemia 11B, BCL-11B, BCL11B/TRDC fusion, COUP-TF-interacting protein 2, CTIP-2, CTIP2ZNF856B, hRit1, hRIT1-alpha, Radiation-induced tumor suppressor gene 1 protein, RIT1, zinc finger protein hRit1 alpha | |
BCL11B | |
IgG1 | |
Protein G purified |
1F8H9 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Apoptosis, Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
64919 | |
Purified recombinant fragment of human BCL11B (AA: 1-150) expressed in E. Coli. | |
Primary | |
95.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title