Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL11B Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600262AF350
Description
BCL11B Polyclonal antibody specifically detects BCL11B in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
BCL11B | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
B-cell CLL/lymphoma 11B, B-cell CLL/lymphoma 11B (zinc finger protein), B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein, B-cell lymphoma/leukaemia 11B, B-cell lymphoma/leukemia 11B, BCL-11B, BCL11B/TRDC fusion, COUP-TF-interacting protein 2, CTIP-2, CTIP2ZNF856B, hRit1, hRIT1-alpha, Radiation-induced tumor suppressor gene 1 protein, RIT1, zinc finger protein hRit1 alpha | |
A synthetic peptide which maps to a region between residues 1 and 50 of human B-cell CLL/lymphoma 11B using the numbering given in Swiss-Prot entry Q9C0K0 (GeneID 64919). | |
0.1 mL | |
Apoptosis, Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
64919 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction