Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2L10 Mouse anti-Human, Clone:8A2F9, Abnova™

Catalog No. 89334407 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-407 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-407 Supplier Abnova Corporation Supplier No. MAB17917
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human BCL2L10.

The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains conserved BH4, BH1 and BH2 domains. This protein can interact with other members of BCL-2 protein family including BCL2, BCL2L1/BCL-X(L), and BAX. Overexpression of this gene has been shown to suppress cell apoptosis possibly through the prevention of cytochrome C release from the mitochondria, and thus activating caspase-3 activation. The mouse counterpart of this protein is found to interact with Apaf1 and forms a protein complex with Caspase 9, which suggests the involvement of this protein in APAF1 and CASPASE 9 related apoptotic pathway. [provided by RefSeq]

Specifications

Antigen BCL2L10
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8A2F9
Conjugate Unconjugated
Description BCL2-like 10 (apoptosis facilitator)
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunofluorescence (1:200-1:1000)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user
Formulation In PBS (0.05% sodium azide)
Gene BCL2L10
Gene Alias BCL-B, Boo, Diva, MGC129810, MGC129811
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 31-186 of human BCL2L10.
Quantity 100 μg
Gene ID (Entrez) 10017
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.