Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCL2L12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554267 Supplier Invitrogen™ Supplier No. PA554267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSESPRPCSL PIRPCYGLEP GPATPDFYAL VAQRLEQLVQ EQLKSPPSPE LQGPPSTEKE AILRRLVALL EEEAEVINQK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.

This gene encodes a member of a family of proteins containing a Bcl-2 homology domain 2 (BH2). The encoded protein is an anti-apoptotic factor that acts as an inhibitor of caspases 3 and 7 in the cytoplasm. In the nucleus, it binds to the p53 tumor suppressor protein, preventing its association with target genes. Overexpression of this gene has been detected in a number of different cancers. There is a pseudogene for this gene on chromosome 3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL2L12
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BCL2L12
Gene Accession No. Q9HB09
Gene Alias 2810475P17Rik; 5430429M05Rik; BCL2 like 12; Bcl-2 related proline-rich protein; Bcl2l12; Bcl2-L12; Bcl2-L-12; BCL2-like 12 (proline rich); bcl-2-like protein 12; Bcl-2-related proline-rich protein; Bcl-L12; BPR
Gene Symbols BCL2L12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BCL2L12. Recombinant protein control fragment (Product #RP-92039).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.