Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCL2L12 Recombinant Rabbit Monoclonal Antibody (4Z2Z8)
GREENER_CHOICE

Catalog No. PIMA538012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538012 Supplier Invitrogen™ Supplier No. MA538012
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: MCF7, PC-3 Immunogen sequence: GPPSTEKEAI LRRLVALLEE EAEVINQKLA SDPALRSKLV RLSSDSFARL VELFCSRDDS SRPSRACPGP PPPSPEPLAR LALAMELSRR VAGLGGTLAG L.

This gene encodes a member of a family of proteins containing a Bcl-2 homology domain 2 (BH2). The encoded protein is an anti-apoptotic factor that acts as an inhibitor of caspases 3 and 7 in the cytoplasm. In the nucleus, it binds to the p53 tumor suppressor protein, preventing its association with target genes. Overexpression of this gene has been detected in a number of different cancers. There is a pseudogene for this gene on chromosome 3. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL2L12
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 4Z2Z8
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BCL2L12
Gene Accession No. Q9HB09
Gene Alias 2810475P17Rik; 5430429M05Rik; BCL2 like 12; Bcl-2 related proline-rich protein; Bcl2l12; Bcl2-L12; Bcl2-L-12; BCL2-like 12 (proline rich); bcl-2-like protein 12; Bcl-2-related proline-rich protein; Bcl-L12; BPR
Gene Symbols BCL2L12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-300 of human BCL2L12 (Q9HB09).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83596
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.