Learn More
Invitrogen™ BCL2L15 Polyclonal Antibody

Description
The antibody detects endogenous levels of total BCL2L15 protein.
Specifications
Specifications
| Antigen | BCL2L15 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | BCL2L15 |
| Gene Accession No. | Q5TBC7 |
| Gene Alias | BCL2 like 15; BCL2L15; bcl2-L-15; BCL2-like 15; bcl-2-like protein 15; BCLl2-like 15; Bfk; C1orf178; Gm566; pro-apoptotic Bcl-2 protein; RGD1562344 |
| Gene Symbols | BCL2L15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.