Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2L2 Mouse anti-Human, Clone:4G12D11, Abnova™

Catalog No. 89334386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-386 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-386 Supplier Abnova Corporation Supplier No. MAB17938
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human BCL2L2.

This gene encodes a member of the BCL-2 protein family. The proteins of this family form hetero- or homodimers and act as anti- and pro-apoptotic regulators. Expression of this gene in cells has been shown to contribute to reduced cell apoptosis under cytotoxic conditions. Studies of the related gene in mice indicated a role in the survival of NGF- and BDNF-dependent neurons. Mutation and knockout studies of the mouse gene demonstrated an essential role in adult spermatogenesis. [provided by RefSeq]

Specifications

Antigen BCL2L2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4G12D11
Conjugate Unconjugated
Description BCL2-like 2
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene BCL2L2
Gene Alias BCL-W, BCLW, KIAA0271
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 6-118 of human BCL2L2.
Quantity 100 μg
Gene ID (Entrez) 599
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.