Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP287068
Description
Bcl3 Polyclonal specifically detects Bcl3 in Human samples. It is validated for Western Blot.Specifications
Bcl3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
B-cell CLL/lymphoma 3, B-cell leukemia/lymphoma 3, B-cell lymphoma 3 protein, B-cell lymphoma 3-encoded protein, BCL-3, BCL4, chronic lymphatic leukemia protein, D19S37, Proto-oncogene BCL3 | |
The immunogen is a synthetic peptide directed towards the middle region of human Bcl3 | |
100 μL | |
Adaptive Immunity, Cell Cycle and Replication, Chromatin Research, Immunology, Signal Transduction, Transcription Factors and Regulators | |
602 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction