Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BCL6 Monoclonal Antibody (BCL-DWN), PE, eBioscience™, Invitrogen™

Catalog No. 12545380
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-545-380 25 μg
50-112-2326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-545-380 Supplier Invitrogen™ Supplier No. 12545380
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This BCL-DWN monoclonal antibody reacts with mouse Bcl-6, which is also known as B-cell lymphoma 6. A member of the POZ family of transcriptional repressors, Bcl-6 is expressed most abundantly in germinal center B cells, as well as in differentiating and activated B cells. This 79-kDa protein is essential for germinal center B cell proliferation and survival. Moreover, during B cell development, Bcl-6 plays an important role in promoting pre-B cell survival to enable the generation of a diverse B cell repertoire. Moreover, Bcl-6 is expressed constitutively in diffuse large B cell lymphomas (DLBCLs), Burkitt's lymphoma, and non-Hodgkin's lymphoma. Reports also suggest expression of Bcl-6 in CD4+ T follicular helper cells and memory T cells. Bcl-6 has been reported to be antagonized by Blimp-1. Applications Reported: This BCL-DWN antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This BCL-DWN antibody has been tested by flow cytometric analysis of splenocytes isolated from sheep red blood cell-immunized mice. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin's lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt's lymphomas and in nodular, lymphocyte predominant Hodgkin's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL6
Applications Flow Cytometry
Classification Monoclonal
Clone BCL-DWN
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene BCL6
Gene Accession No. P41183
Gene Alias B cell CLL/lymphoma 6; B cell leukemia/lymphoma 6; B-cell CLL/lymphoma 6; B-cell CLL/lymphoma 6 (zinc finger protein 51); B-cell leukemia/lymphoma 5; B-cell leukemia/lymphoma 6; B-cell lymphoma 5 protein; B-cell lymphoma 5 protein (BCL5); B-cell lymphoma 6 protein; B-cell lymphoma 6 protein homolog; B-cell lymphoma 6 protein transcript; BCL5; BCL-5; Bcl6; Bcl-6; BCL6 transcription repressor; BCL6A; cys his2 zinc finger transcription factor; cys-his2 zinc finger transcription factor; LAZ3; LAZ-3 protein; Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3); lymphoma-associated zinc finger gene on chromosome 3; protein LAZ-3; ZBTB27; zinc finger and BTB domain-containing protein 27; Zinc finger and BTB domain-containing protein 27 (ZBTB27); Zinc finger protein 51; Zinc Finger Protein 51 (ZNF51); zinc finger transcription factor BCL6S; ZNF51
Gene Symbols BCL6
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12053
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.