Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL6 Monoclonal Antibody (OTI8A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA804243

Description
BCL6 is a zinc finger transcription factor and contains an N-terminal POZ domain. This protein acts as a sequence-specific repressor of transcription, and has been shown to modulate the transcription of START-dependent IL-4 responses of B cells. This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors.Specifications
BCL6 | |
Monoclonal | |
Unconjugated | |
BCL6 | |
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51 | |
Mouse | |
Affinity Chromatography | |
RUO | |
604 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
OTI8A3 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P41182 | |
BCL6 | |
Human recombit protein fragment corresponding to amino acids 1-300 of human BCL6 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction