Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL6 Mouse anti-Human, Biotin, Clone: 1E6B1, Proteintech
SDP

Catalog No. 501737229 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7229 Supplier Proteintech Group Inc Supplier No. BIOTIN66340100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody only recognize human species.

BCL6 is a zinc finger transcription factor and contains an N-terminal POZ domain. This protein acts as a sequence-specific repressor of transcription, and has been shown to modulate the transcription of START-dependent IL-4 responses of B cells. This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors.

Specifications

Antigen BCL6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1E6B1
Concentration 0.67 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BCL6
Gene Accession No. P41182
Gene Alias B cell CLL/lymphoma 6, B cell lymphoma 5 protein, B cell lymphoma 6 protein, BCL6, LAZ3, Protein LAZ 3, ZBTB27, Zinc finger protein 51, ZNF51
Gene Symbols BCL6
Host Species Mouse
Immunogen BCL6 Fusion Protein Ag15519
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 604
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.