Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL6 Mouse anti-Human, Clone: BCL6/1475, Abnova™

Catalog No. 89034989 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-989 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-989 Supplier Abnova Corporation Supplier No. MAB14969
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human BCL6.

  • High specificity
  • High reactivity

Specifications

Antigen BCL6
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone BCL6/1475
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human BCL6.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Accession No. P41182
Gene Alias BCL5/BCL6A/LAZ3/ZBTB27/ZNF51
Gene Symbols BCL6
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 256-389 of human BCL6.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 604
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.