Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL6 Rabbit anti-Human, Unconjugated, Clone: BCL6/1951R, Abnova™

Catalog No. 89339747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-747 Supplier Abnova Corporation Supplier No. RAB00708
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

BCL6 Monoclonal antibody specifically detects BCL6 in Human samples. It is validated for IHC-P.

Specifications

Antigen BCL6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BCL6/1951R
Conjugate Unconjugated
Description B-cell CLL/lymphoma 6
Gene BCL6
Gene Alias BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 256-389 of human BCL6
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 604
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.