Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BCL6B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563671
Description
Immunogen sequence: RLSPTAATVQ FKCGAPASTP YLLTSQAQDT SGSPSERARP LPGSEFFSCQ NCEAVAGCSS GLDSLVPGDE D Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 61%.
BCL6B acts as a sequence-specific transcriptional repressor in association with BCL6. BCL6B may function in a narrow stage or be related to some events in the early B-cell development.
Specifications
BCL6B | |
Polyclonal | |
Unconjugated | |
BCL6B | |
B cell CLL/lymphoma 6, member B; B cell CLL/lymphoma 6B; BAZF; B-cell CLL/lymphoma 6 member B protein; B-cell CLL/lymphoma 6, member B; B-cell CLL/lymphoma 6, member B (zinc finger protein); B-cell CLL/lymphoma 6B; Bcl6-associated zinc finger protein; Bcl6b; BCL6B transcription repressor; BCL6B, transcription repressor; LOW QUALITY PROTEIN: B-cell CLL/lymphoma 6 member B protein; RGD1563179; ZBTB28; zinc finger protein 62; ZNF62 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
255877 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N143 | |
BCL6B | |
Recombinant protein corresponding to Human BCL6B. Recombinant protein control fragment (Product #RP-101832). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction